Learn More
StatLab™ MasterTech Iron Stain Kit
SDP
Catalog No. STLKTIROPT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTIROPT 1 pt.
STLKTIRO 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTIROPT Supplier StatLab™ Supplier No. KTIROPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Iron Stain Kit stains ferric iron in under 30 minutes.
  • A Working Prussian Blue Solution serves as the primary stain, producing blue ferric iron against a pink to red background with red nuclei, counterstained by Nuclear Fast Red
  • No filtration required when using MasterTech Nuclear Fast Red
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit