Learn More
StatLab™ MasterTech Light Green Counterstain
SDP
Catalog No. STLSTLGCLT
Change view
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
1 pt.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSTLGCLT 1 L
STLSTLGCPT 1 pt.
STLSTLGCGAL 1 gal./Bottle, 4 Bottles/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSTLGCLT Supplier StatLab™ Supplier No. STLGCLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Used for the staining of collagen fibers and especially for Masson's trichrome staining.
  • Used for routine staining methods in histology
  • Used in the MasterTech AFB, Fites, GMS and PAS stain kits
Specifications
Quantity 1 L
Product Type Light Green