Learn More
StatLab MasterTech Masson's Trichrome Stain Kit
SDP
Catalog No. 21033403
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-403 100 mL
21-033-404 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-403 Supplier StatLab Supplier No. KTMTR2
Add to Cart
Add to Cart
Stains for collagen and muscle in as little 20 minutes
  • Trichrome stains are commonly used to identify increases in collagenous tissue to assist in the diagnosis of diseases such as liver cirrhosis
  • This kit includes a 20 minute microwave procedure and the traditional one hour and forty-five minute procedure
  • Biebrich Scarlet/Acid Fuchsin and Aniline Blue serve as the primary stains, producing red cytoplasm, intercellular fiber, keratin, and muscle, blue collagen and mucus with purple to black nuclei, counterstained by Weigerts Hematoxylin
  • 100 mL Stain Kit yields between 80-100 slides
  • 100 mL Kit Consists of: Bouins Fluid 100 mL, Biebrich Scarlet/Acid Fucshin 100 mL, Phosphomolybdic/Phoshotungstic Acid 100 mL, Aniline Blue Stain 100 mL, 1% Acetic Acid 100 mL and Weigerts AandB Hematoxylin 100 mL each
  • A MasterTech Special Stain Kit
Specifications
Quantity 100 mL
Product Type Masson's Trichrome