Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ MasterTech Nuclear Fast Red Stain
SDP

Catalog No. STLSTNFR100
Change view
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
1 pt.
100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSTNFR100 100 mL
STLSTNFRPT 1 pt.
STLSTNFRLT 1 L
STLSTNFRGAL 1 gal./Bottle, 4 Bottles/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSTNFR100 Supplier StatLab™ Supplier No. STNFR100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Nuclear Fast Red Stain is a highly versatile reagent that selectively stains nuclear chromatin red, while the nonspecific signal is detected in pink tones.

  • For the staining of the nuclei where it will react with negatively charged components such as nucleic acids in the nucleus
  • An ideal alternative to hematoxylin which would otherwise obscure the substrate colors

Specifications

Quantity 100 mL
Product Type Nuclear Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.