Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Oil Red O Stain Kit
SDP

Catalog No. 21033405
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-405 100 mL
21-033-406 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-405 Supplier StatLab Supplier No. KTORO
Add to Cart
Add to Cart

Stains fat and neutral fats in 10 minutes

  • Oil Red O serves as the primary stain, producing red fat with blue nuclei stained by the Hematoxylin
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 100 mL Kit Consists of: Propylene Glycol Pint, Oil Red Stain O 100 mL and Modified Mayers Hematoxylin 100 mL
  • A MasterTech Special Stain Kit

Specifications

CAS 64-19-7, 57-55-6
Grade Laboratory
Color Red
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.