Learn More
StatLab MasterTech Phosphomolybdic Acid
SDP
Catalog No. 23640018
Change view
Click to view available options
Quantity:
125 Pk.
25 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-018 25 Pk.
23-640-017 125 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-018 Supplier StatLab Supplier No. AHPPMA25
Harmful if swallowed, in contact with skin, or if inhaled
A reagent used in histological staining applications.
  • Component of the Masson's Trichrome stain kit used in histology laboratories.
  • Essential for differentiating collagen from smooth muscle in tissue samples for anatomical and pathological evaluation.
Specifications
Quantity 25 Pk.
Product Type Phosphotungstic/Phosphomolybdic Acid