Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Special Stain Kit, 12% Acetic Acid
SDP

Catalog No. 23640182
Change view
Click to view available options
Color:
Colorless to Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-640-182 Colorless to Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-182 Supplier StatLab Supplier No. KC0343
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

12% Acetic Acid

  • 12% Acetic Acid is a component for the MasterTech Colloidal Iron Stain Kit
  • An acetic acid bath removes excess blue and reveals the different tissue elements whose color varies depending on the molecules fixed by the dye

Specifications

Physical Form Aqueous Solution
Color Colorless to Yellow
Quantity 1 L
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.