Learn More
StatLab MasterTech Special Stain Kit Component, One Step Trichrome Stain (Blue and Red)
SDP
Catalog No. 23640264
Change view
Click to view available options
Quantity:
1 L
1 p.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-264 1 L
23-640-265 1 p.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-264 Supplier StatLab Supplier No. STOSTBLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
One Step Trichrome Stain (Blue and Red)
  • A MasterTech Special Stain Kit Component
  • One Step Trichrome Stain (Blue and Red) is used in histology lab applications to visualize tissue architecture by providing distinct color contrasts
  • It typically highlights collagen (blue or green), muscle fibers (red), and nuclei (dark)
Specifications
Quantity 1 L
Product Type Stain Kit