Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matching Fluid K, USP - for Color and Achromicity, Ricca Chemical
SDP

Catalog No. R4621011500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4621011500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R4621011500 Supplier Ricca Chemical Company Supplier No. 4621.K16
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Matching Fluid K
Packaging Amber Glass Bottle
Name Note USP, For Color and Achromicity
CAS 10025-77-1, 7647-01-0, 7732-18-5, 7791-13-1
Odor Odorless
Physical Form Liquid
Color Brown/Yellow
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) Water Analyzer
Solubility Infinitely soluble in water
Solution Type Calibration Solution
Type Color Reference Standard
Solvent or Matrix Water
Quantity 500 mL
Grade USP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.