Learn More
Brady™ Material Handling, Shipping & Receiving Labels - Self-Sticking Polyester
Catalog No. 19110985 Shop All Brady Corporation Products
Change view
Click to view available options
Display Text:
DNGR.CHOCK
HAZARDOUS WASTE
USED OIL
WASTE FLAMMABLE
WASTE OIL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
19-110-985 HAZARDOUS WASTE
19-110-988 USED OIL
19-111-359 DNGR.CHOCK
19-110-990 WASTE OIL
19-110-989 WASTE FLAMMABLE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-110-985 Supplier Brady™ Supplier No. 49896
Only null left
Add to Cart
Add to Cart
Self-Sticking Polyester (B-302) is a flexible polymer plastic over-laminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals.
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (up to 230°F)
  • Flexible material conforms to most surfaces while protected graphics hold up to harsh environments, abrasion, and repeated spills and cleanings
  • Permanent, pressure sensitive adhesive forms a long-lasting bond to even the most difficult surfaces and shapes
  • Cold temperature adhesive allows the signage to be applied at 0°F (-18°C)
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Description Label
Adhesive Type Permanent Acrylic, Self-adhesive
Finish Gloss
Format Label
Material Polyester
Printable Sides Single-side Printable
Product Type Label
Display Text HAZARDOUS WASTE
Languages English
Quantity 5/Pk.
Shape Rectangle
Width (Metric) 92.71 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov