Learn More
CPC Scientific H-Trp-His-Trp-Leu-Gln-Leu-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific MATE002B
Encompass
ONE-LETTER SEQUENCE: WHWLQLMOLECULAR FORMULA: C45H59N11O8MOLECULAR WEIGHT:882.03STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [65418-88-4], RESEARCH AREA: Hormonal
Catalog No. 50-210-9704
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.