Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrin 3 Rabbit anti-Human, Mouse, Rat, Clone: 10O1D1, R&D Systems™
SDP

Catalog No. NB165711
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165711 20 μg
NB165710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB165711 Supplier R&D Systems Supplier No. NBP31530720UL

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

Matrin 3 Monoclonal specifically detects Matrin 3 in Human∣Mouse∣Rat samples. It is validated for Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Paraffin)∣Western Blot.

Specifications

Antigen Matrin 3
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 10O1D1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias distal 2, DKFZp686K0542, DKFZp686K23100, matrin 3, matrin-3, vocal cord and pharyngeal weakness with distal myopathy
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 700-800 of human Matrin 3 (P43243).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 9782
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.