Learn More
Thermo Scientific™ Matrix™ Blank and Alphanumeric Storage Tubes
GREENER_CHOICE
Catalog No. p-4526110
Change view
Click to view available options
Product Type:
Alphanumeric Storage Tube
Blank Storage Tube
Packaging:
10 Latch Racks of 96 Tubes
10 Refill Carriers of 96 Tubes
10 Snap Racks of 96 Tubes
Bulk, 1000 Tubes
Sterility:
Non-sterile
Sterile
Fit for Purpose:
PCR Ready
Sterile and PCR Ready
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Sterility Fit for Purpose
14-754-662 Alphanumeric Storage Tube 10 Refill Carriers of 96 Tubes Non-sterile PCR Ready
14-754-651 Blank Storage Tube Bulk, 1000 Tubes Non-sterile PCR Ready
14-754-652 Alphanumeric Storage Tube 10 Snap Racks of 96 Tubes Non-sterile PCR Ready
14-754-659 Blank Storage Tube 10 Refill Carriers of 96 Tubes Sterile Sterile and PCR Ready
14-754-664 Blank Storage Tube 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready
14-754-655 Blank Storage Tube Bulk, 1000 Tubes Non-sterile PCR Ready
14-754-665 Alphanumeric Storage Tube 10 Latch Racks of 96 Tubes Non-sterile Sterile and PCR Ready
14-754-656 Blank Storage Tube 10 Latch Racks of 96 Tubes Non-sterile PCR Ready
14-754-658 Blank Storage Tube 10 Refill Carriers of 96 Tubes Non-sterile PCR Ready
14-754-666 Alphanumeric Storage Tube 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready
14-754-657 Blank Storage Tube 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready
14-754-660 Alphanumeric Storage Tube 10 Latch Racks of 96 Tubes Non-sterile PCR Ready
14-754-663 Blank Storage Tube 10 Latch Racks of 96 Tubes Non-sterile PCR Ready
14-754-661 Alphanumeric Storage Tube 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready
14-754-653 Alphanumeric Storage Tube 10 Latch Racks of 96 Tubes Sterile Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 14-754-662 Supplier Thermo Scientific™ Supplier No. 4253
Only null left
Add to Cart
Add to Cart
Store valuable samples where 2D tracking is not required with high-density blank and alphanumeric storage tubes.
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or DeepWell™ block sample storage.
Features:
Highest Quality
  • Matrix storage tubes are manufactured from medical grade virgin polypropylene
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Superior Storage Rack Design
  • Unlike traditional storage tubes or blocks, Thermo Scientific Matrix blank and alphanumeric storage tubes are available in our specially designed, stackable, microplate-footprint Latch Racks of 96 tubes for individual sample, access and removal
  • Enables multichannel pipette access and eliminates contamination risk with a lid that never touches the bench top when open
  • Compatible with automated liquid handling, as the lid can be lifted robotically for high-throughput applications
Clear Designations
  • Clearly marked with high contrast printing on the base of each tube, designating their appropriate well location within a rack, for easy identification
  • Enhance ease of visual identification with colored SepraSeal capping options or labelling on the side of the tube using the ID Scribe instrument
Capping Options
  • Several sealing options available, including the Thermo Scientific Matrix line of CapMats and CapStrips for sealing multiple tubes and the Thermo Scientific Matrix SepraSeal and DuraSeal capping systems for individual tube sealing, access and piercing
  • Applied with the Thermo Scientific SuperSealer, sealing is secure and consistent every time
Compatible with:
Capping systems, SepraSeals, Duraseals, SuperSealer, ID Scribe
Recommended for:
Individual sample retrieval; Shipping samples; Storage of DNA, compounds or any other samples
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1.4 mL
Material Polypropylene
Packaging 10 Refill Carriers of 96 Tubes
Product Line Matrix
Sterility Non-sterile
Capacity (English) 0.0473 fl oz.
Dnase Free Yes
Fit for Purpose PCR Ready
Product Type Alphanumeric Storage Tube