Learn More
Specifications
Specifications
| Chemical Name or Material | Matrix Modifier |
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 0.5% Pd,5000ppm Pd |
| CAS Min % | 88.0, 9.29, 0.35, 0.06 |
| CAS Max % | 92.0, 9.67, 0.36, 0.06 |
| CAS | 7697-37-2, 7440-05-3, 7647-01-0 |
| Physical Form | Liquid |
| Color | Brown |
| Melting Point | Approximately 0°C |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
