Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Matrix™ Reagent Reservoirs
GREENER_CHOICE

Catalog No. 14387067
Change view
Click to view available options
Capacity (Metric):
25 mL
75 mL
100 mL
Material:
Polypropylene
Polystyrene
Packaging:
10 Bags of 10
50 Individually wrapped
Type:
Reagent Reservoir
Reagent Reservoir with Divider
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Material Packaging Type
14-387-067 100 mL Polystyrene 10 Bags of 10 Reagent Reservoir
14-387-065 75 mL Polypropylene - Reagent Reservoir
14-387-068 100 mL Polystyrene 50 Individually wrapped Reagent Reservoir
14-387-069 25 mL Polystyrene 10 Bags of 10 Reagent Reservoir
14-387-070 25 mL Polystyrene 50 Individually wrapped Reagent Reservoir
14-387-071 25 mL Polystyrene 10 Bags of 10 Reagent Reservoir with Divider
14-387-072 25 mL Polystyrene 50 Individually wrapped Reagent Reservoir with Divider
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-387-067 Supplier Thermo Scientific™ Supplier No. 8085
Add to Cart
Add to Cart

Accommodate a variety of applications with these uniquely designed, innovative reagent reservoirs.

Accomodate a variety of applications with uniquely designed, innovative Thermo Scientific™ Matrix Reagent Reservoirs. These sterile reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills. The 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizing recovery if using only a single channel and a small amount of reagent.

Features: 

  • Trough within a trough maximizes amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap-and-tear bags, maintaining sterility while being easy to open
TRUSTED_SUSTAINABILITY

Specifications

Color White
Capacity (Metric) 100 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Packaging 10 Bags of 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.