Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell North™ Matterhorn A89 HDPE Cap Style Hard Hat with 4-Point Ratchet Suspension

Catalog No. 19085629 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Suspension Type:
4-Point Ratchet Suspension
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Suspension Type
19-085-629 4-Point Ratchet Suspension
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-085-629 Supplier Honeywell North™ Supplier No. A89R010000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An all purpose lateral impact HDPE hard hat with foam liner, rain trough, and accessory slots. Includes 4-point nylon ratchet suspension with 3-level height adjustment.

  • Front brim reduces glare and helps shade the eyes and face in bright sunlight
  • Type 2 to provide protection against vertical and angled impacts up to 60°
  • Additional padding generally makes these hard hats heavier and more bulky than Type 1
  • Rated Class E for high-voltage electrical protection up to 20 kV
  • Should be worn when indicated by OSHA or where swinging objects (such as hooks or chains) are a concern

Specifications

Certifications/Compliance Meets ANSI Z89.1-2009 and CSA Z94.1-2005 Type 2 Class E
Color White
For Use With (Application) Head Protection
Includes Accessory Slot, Foam Liner, and Rain Trough
Material Polyethylene
Product Type Hard Hat
Size Universal
Suspension Type 4-Point Ratchet Suspension
Ventilated Not Ventilated
Head Style Short Brim (Cap Style)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.