Learn More
ProteinSimple Maurice CE-SDS Running Buffer - Bottom
Supplier:  ProteinSimple 046385
Encompass
Running buffer for CE-SDS application. 15 mL. For use with CE-SDS cartridge (PS-MC02-S) on Maurice (090-000) and Maurice S. (090-001) systems only.
Catalog No. NC1233407