Learn More
R&D Systems Maurice CE-SDS Running Buffer - Top, 12/pk
SDP
Catalog No. NC1233405 Shop All Novus Biologicals Products
Encompass
Pre-filled vial assembly containing top running buffer for CE-SDS application. For use with the cartridge insert (046-124) of the CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only. 12 Vials.
Catalog No. NC1233405 Supplier R&D Systems Supplier No. 046384/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
Pre-filled vial assembly containing top running buffer for CE-SDS application. For use with the cartridge insert (046-124) of the CE-SDS cartridge (PS-MC02-S) on Maurice and Maurice S. systems only. 12 Vials.