Learn More
Novus Biologicals Maurice cIEF Chemical Test Kit (8)
Supplier: Novus Biologicals 046036
The cIEF Chemical Test Kit is designed to confirm the overall performance of the Maurice and Maurice C. systems with the cIEF cartridge (PS-MC02-C) on 8 separate occasions. The kit contains Anolyte, Catholyte, Methyl Cellulose, Fluorescence Calibration Standard and a System Suitabilty kit. Each time, the performance can be checked up to 24 hours. The expiration date for this kit is 12 months from date of manufacture. 8 runs/kit.