Learn More
R&D Systems Maurice cIEF System Suitability Kit
SDP
Catalog No. NC1197043 Shop All Novus Products
Encompass
The cIEF System Suitability Kit is used to run a control test for cIEF application on Maurice and Maurice C. systems on 8 separate occasions. Each time, the performance can be checked up to 24 hours. The kit contains a vial of lyophilized Peptide Panel and a vial of System Suitability Test mix. 8 runs/kit. The expiration date for this kit is 12 months from date of manufacture.
Catalog No. NC1197043 Supplier R&D Systems Supplier No. 046044/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
The cIEF System Suitability Kit is used to run a control test for cIEF application on Maurice and Maurice C. systems on 8 separate occasions. Each time, the performance can be checked up to 24 hours. The kit contains a vial of lyophilized Peptide Panel and a vial of System Suitability Test mix. 8 runs/kit. The expiration date for this kit is 12 months from date of manufacture.