Learn More
Novus Biologicals Maurice Glass Reagent Vials, 2 mL, 100/pk
Supplier: Novus Biologicals 046017
Screw-top glass vials, 2 mL. 100/pk. For use with Maurice, Maurice S., and Maurice C. systems only.