Learn More
Medchemexpress LLC Mauritianin | 109008-28-8 | MFCD32197360 | 99.8% | 740.66 g·mol⁻¹ | C33H40O19 | 10 MG
Supplier: Medchemexpress LLC HYN503810MG
Mauritianin is an orally active kaempferol glycoside isolated from the flowers and leaves of Acalypha indica. It is reported to inhibit topoisomerase I and is provided as a high-purity research reagent with supporting product documentation and safety data.
- Orally active kaempferol glycoside with reported biological activity.
- High purity: 99.8% as listed by the supplier.
- Chemical formula C33H40O19 and molecular weight 740.66 g·mol⁻¹.
- Supplied as a solid in small research quantities (example: 10 mg).
- Product data sheet and safety data sheet available for handling and characterization.
- Suitable for research into flavonoid bioactivity and topoisomerase-related mechanisms.