Learn More
Ansell KleenGuard™ Maverick™ Safety Glasses
GREENER_CHOICE
Catalog No. 19005090 Shop All Ansell Products
Change view
Click to view available options
Frame Color:
Black
Black, Orange Tips
Clear, Orange Tips
Lens Tint:
Clear
Smoke
Lens Coating:
Anti-fog
Anti-reflective
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-005-090 Black Clear Anti-fog
19-005-091 Black Smoke Anti-fog
19-005-092 Black, Orange Tips Clear Anti-reflective
19-005-093 Clear, Orange Tips Clear Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-005-090 Supplier Ansell Supplier No. 49309
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Maverick™ Safety Glasses are designed to enable rather than inhibit, which allows for maximum productivity and safety for you and your employees.
KleenGuard Maverick Safety Eyewear's stylish good looks and durable, lightweight construction, you don't have to sacrifice looking good to be safe. Premium anti-fog and anti-scratch coating that passes the EN166 'N' mark test for resistance to fogging and EN166 'K' mark test for resistance to surface damage by fine particles.
Maverick Eye Protection provides 99.9% UVA/UVB/UVC protection. They also come with comfort touch points on the temples and nose bridge. The eyewear meets the optical requirements and impact protector requirements of the ANSI Z87.1+ standards, with integrated side shields and browguard to help the user feel confident they are protected.
Both men and women will find the stylish KleenGuard Maverick Safety Eyewear to be comfortable and functional, giving you and your employees uncompromised protection.
  • Features a modern sleek and stylish lightweight design that your employees will be happy to wear
  • Meets the ANSI Z87.1+ standards for optical and impact protector requirements
  • Polycarbonate lenses provide 99.9% UVA/UVB/UVC protection
  • Premium Anti-Fog and Premium Anti-Scratch lens coatings for better performance and longer life
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1+, EN166 (K,N)
Description Universal Size
Lens Tint Clear
Frame Color Black
No. per Case 12/Cs.
No. per Pack 1 Pair
Lens Material Polycarbonate
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.