Learn More
DIVERSIFIED AIR SYSTEMS MAXFLO PC-1200
Supplier:  DIVERSIFIED AIR SYSTEMS MAXFLOPC1200
Encompass
MAXFLO PC-1200 with (1) 6" dia. x 10' long fumeextractor arm, nominal 1200 CFM, manual yellow pushbutton pulse-jet filter cleaning system with minihelic filtercleaning gauge, dust drawer, 1.5 HP TEFC motor/blower,120/1/60 with 25' yellow power cord with power cordhook, on/off switch, direct drive backward inclined airfoilnon-overloading blower, large 295 sq/ft fire retardantNANO MERV 15 cartridge filter, internal baffle protectingcartridge filter from sparks, 10 inch rear wheels and 5inch front swivel casters, grab handle, textured powdercoat finish, 76 dBA.
Catalog No. NC2320309
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.