Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiDry™ Zero™ Double-Dipped MicroFoam Nitrile-Coated Gloves with Thermal Lining

Catalog No. 19046440 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-440 2X-Large
19-046-436 Small
19-046-438 Large
19-046-439 X-Large
19-046-437 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-046-440 Supplier PIP™ Supplier No. 56451/XXL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

THERMtech™ technology platform offers thermal resistance up to -10°C (14°F) inside the glove under high activity, with a coating designed to remain flexible for temperature up to -30°C (-22°F).

  • Seamless construction increases comfort and breathability
  • MicroFoam Nitrile coating is compatible with light oils to enhance grip and abrasion resistance
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Liquid repellence keeps moisture away from the skin to ensure hands remain warm
  • High degree of thermal insulation without sacrificing flexibility, dexterity, weight, and comfort
  • Gloves help to prevent the effects of wind chills for outdoor applications
  • Coating remains flexible even in extremely cold environments
  • Optimized grip delivered though the ultra soft coating to ensure maximum flex and contact with the items being handled
  • Insulation liner is bonded away from flex and movement points of the hand (i.e. knuckles) to ensure comfort
  • All ingredients used in manufacturing and construction are compliant with the REACH legislation
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Silicone-free

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Coating Material Nitrile
Color Purple
Exterior Finish Fully Textured
Hand Specific
Material Nylon Lycra
Product Type Cold-resistant Gloves
Size 2X-Large
For Use With (Application) Outdoor Construction/Maintenance, Agriculture, Cold Food Storage, and Chilled Distribution
Glove Cuff Style Knit
Liner Material Thermal
Series 56-451
Yarn Gauge 15
Length (English) 11.4 in.
Length (Metric) 29 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.