Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ MaxiFlex™ Endurance™ MicroFoam Nitrile Coated Nylon Gloves with Micro Dot Palm

Catalog No. 191301179E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-1179E X-Large
19-130-1179B Small
19-130-1179C Medium
19-130-1179D Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1179E Supplier PIP™ Supplier No. 34844/XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Seamless knit gloves incorporate raised micro dots to increase durability, and offer additional cushioning in highly repetitive applications that require precision handling.

  • MicroFoam Nitrile coating is compatible with light oils to enhance grip and abrasion resistance
  • Raised micro dots increase durability and offer more cushioning in highly repetitive applications that require precision handling
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Patented micro-foam nitrile coating offers 360° breathability
  • 25% thinner than most foam nitrile gloves while offering twice the mechanical performance
  • Form, fit and feel mimics the hand at rest to reduce hand fatigue and increase comfort
  • Latest knitting technology is utilized to produce smooth and rounded fingertips, improving fingertip sensitivity
  • Consistent liner penetration from the nitrile leaves only the soft liner resting against the skin
  • Micro-cup finish allows for a controlled, optimized grip
  • All ingredients used in the manufacturing and construction are compliant with the REACH legislation
  • Gloves are laundered prior to final packaging to ensure they are clean and safe for use
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • 100% Silicone free

Specifications

ANSI Abrasion Rating ANSI 3
Construction Seamless Knit
Coating Material Nitrile MicroFoam
Color Gray
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
Style Standard
For Use With (Application) General Assembly, Micro-Engineering, Automotive Components, Painting, and Horticulture
Glove Cuff Style Knit
Series 34-844
Yarn Gauge 15
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.