Learn More
3M™ Maxim™ Elite 1000 Series Safety Glasses
Catalog No. 19072202 Shop All 3M Products
Change view
Click to view available options
Lens Tint:
Blue, Mirror
Brown
Clear
Gray
Light Gray
Photochromic
Purple, Mirror
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-072-202 Blue, Mirror
19-072-195 Purple, Mirror
19-072-203 Clear
19-072-201 Light Gray
19-072-200 Gray
19-072-198 Brown
19-072-196 Photochromic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-072-202 Supplier 3M™ Supplier No. 7100222267
This item is not returnable. View return policy
3M™ Maxim™ Elite 1000 Series Safety Glasses with ANSI Z87.1-2015 safety compliance
  • The next generation Maxim design offers upgraded features with perfomance lens technology
  • Influenced by lifestyle eyewear with stylish wrap-around lens design
  • Lightweight rubber painted matte frame material with embossed 3M logo
  • Blue or blue-violet mirrored lens options increase sunlight reflection, perfect for working in bright conditions
  • 3M eye protection meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015
Description Model: MXE1008AF-CLR
Lens Tint Blue, Mirror
Product Type Safety Glasses
For Use With (Application) Primary Metals, Chemicals, Automotive, Masonary, UV Protection, Grinding, Facility Maintenance, Construction, Sub Applications, Transportation, Assembly, Remediation, Machine Operations, Law Enforcement, Chemical Handling, Pharmaceuticals, Machining, Mining, Riveting, Food and Beverage, Drilling, Food and Beverage Manufacturing, Aviation, Pouring/Casting, Chiseling, Sawing, General Manufacturing, Power Fastening, Oil and Gas, Demolition
Frame Color Clear
No. per Case 20/Cs.
Temple Style Classic
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Frame Type Full Frame