Learn More
Thermo Scientific™ Maxima First Strand cDNA Synthesis Kit Reaction Mix (5X)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific Maxima 5X Reaction Mix is a component of the Maxima First Strand cDNA Synthesis Kits for RT-qPCR (K1671/K1672/K1641/K1642) made available here for separate purchase. The 5X Reaction Mix is comprised of reaction buffer, dNTPs, and oligo (dT)18 and random hexamer primers.
Oligo (dT)18 is single-stranded deoxythymine sequence used for priming reactions catalysed by reverse transcriptase.
Related products
Maxima First Strand cDNA Synthesis Kit for RT-qPCR, with dsDNase (Cat. No. K1671)
Maxima First Strand cDNA Synthesis Kit for RT-qPCR (Cat. No. K1641)
Specifications
Specifications
| Concentration | 5X |
| Content And Storage | • 5X Reaction Mix with oligo(dT)18 and random hexamer primers |
| Format | Kit |
| For Use With | Maxima First Strand cDNA Synthesis Kit for RT-qPCR |
| Quantity | 0.8 mL |
| Shipping Condition | Dry Ice |
| Final Product Type | First-Strand cDNA |
| No. of Reactions | 200 Reactions |
| Reaction Format | Premixed Components |
| Reagent Type | Reverse Transcription |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.