Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Maxitips - Pipette Tips
GSA_VA

Catalog No. 2137952 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
1 to 10 mL
2.5 to 10 mL
Quantity:
100 Pk.
200 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
21-379-52 2.5 to 10 mL 200 Pk.
21-379-59 1 to 10 mL 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-379-52 Supplier Eppendorf™ Supplier No. 022291351
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Maxitip S requires 21-379-53 Barrel/Piston or 21-379-51 Barrel/Piston set—order separately.

Eppendorf Maxitips are designed for large volume liquid handling, offering high precision and compatibility with Eppendorf pipetting systems. Ideal for diverse lab applications, they ensure accuracy and ease of use, enhancing workflow efficiency.

  • Use Maxitips P to aspirate 1 mL to 10 mL from beakers using the positive displacement principle
  • Use Maxitips S in combination with the Eppendorf Maxitip to aspirate between 2.5 mL and 10 mL liquid from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips ensures accurate dispensing of non-aqueous liquids

Order Info

Valves are also available for Maxitip S™ and Maxitip G tips to eliminate dripping caused by reagents with high vapor pressure (see 21-379-54). Maxitip P: Purchase valves and tips separately.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2.5 to 10 mL
Filtered Non-filtered
Sterility Non-sterile
Model Maxitip S (air)
Length (English) 6.75 in.
Length (Metric) 17 cm
Tip Style Specialty
Quantity 200 Pk.
Product Type Pipette Tips
For Use With (Equipment) Positive Displacement System
Graduated Yes
Product Line Maxitip
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.