Learn More
Fluidigm Corp MAXPAR X8 ANTIBODY LABELING166
SDP
Supplier:  Fluidigm Corp 201166A
Encompass_Preferred
This Maxpar kit lets you conjugate IgG antibodies with the 166Er metal isotope, with each kit containing enough material to conjugate 440 antibodies in 100 g amounts.
Catalog No. NC0482384
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.