Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Catalog No. 14512319
Change view
Click to view available options
Product Type:
Adhesive Mat applicator
Dual Height Adjustable Clamp
Flask Clamp
Flask Clamp Kit
Media Bottle Clamp
Microplate/Deep-Well Plate Clamp
Separatory Funnel Clamp
Vessel Clamp
Holds:
1 L Erlenmeyer Flask
10 mL Erlenmeyer Flask
1000 mL Square Media Bottle
125 mL Erlenmeyer Flask
2 L Erlenmeyer Flask
25 mL Erlenmeyer Flask
250 mL Erlenmeyer Flask
2500 mL Low-form Culture Flask
2800 mL Fernbach Flask
300 mL Erlenmeyer Flask
4 L Erlenmeyer Flask
5 L Erlenmeyer Flask
50 mL Erlenmeyer Flask
500 mL Erlenmeyer Flask
500 mL Square Media Bottle
Flasks
Microplate/Deep-Well Plate
Separatory Funnel
Single Vessel
Vessel
Vessels
For Use With (Equipment):
Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds For Use With (Equipment)
14-512-319 Flask Clamp 2500 mL Low-form Culture Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
11-675-475 Flask Clamp Kit Flasks Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-315 Flask Clamp 10 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
22-262-769 Flask Clamp 25 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
11-675-476 Flask Clamp 50 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-272-4AA Flask Clamp 125 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-272-4B Flask Clamp 250 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-316 Flask Clamp 300 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
22-256-959 Flask Clamp 500 mL Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-272-4F Flask Clamp 1 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
22-262-773 Flask Clamp 2 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-317 Flask Clamp 4 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-318 Flask Clamp 5 L Erlenmeyer Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-320 Flask Clamp 2800 mL Fernbach Flask Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
14-512-314 Microplate/Deep-Well Plate Clamp Microplate/Deep-Well Plate Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
01-258-056 Vessel Clamp Single Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
01-258-057 Dual Height Adjustable Clamp Vessel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
01-258-062 Media Bottle Clamp 500 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
01-258-063 Media Bottle Clamp 1000 mL Square Media Bottle Solaris Open Air and Temperature Controlled Shakers, MaxQ 6000 Shakers, MaxQ 416 HP Shakers
01-258-079 Separatory Funnel Clamp Separatory Funnel Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
01-258-078 Adhesive Mat applicator Vessels Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options
Catalog No. 14-512-319 Supplier Thermo Scientific™ Supplier No. 30161
Add to Cart
Add to Cart
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Clamps are sold as individual units.
TRUSTED_SUSTAINABILITY
Specifications
Holds 2500 mL Low-form Culture Flask
Product Type Flask Clamp
For Use With (Equipment) Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers