Learn More
Description
Includes
CD ROM, which contains the Firmware, installation software and Technical Manual.
- Modify or Create Automated Methods for Your Maxwell(R) 16 Instrument
- Optimize performance for your sample type and assay
- Control key operating parameters
- Simple interface makes programming easy
Order Info
Flexi Method Firmware ordered with the purchase of a new AS2000 Instrument will be installed at the factory.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Description | Maxwell 16 Flexi Method firmware on CD |
| Isolation Technology | Magnetic Bead |
| For Use With (Equipment) |
|
| For Use With (Application) | DNA purification, RNA purification, recombinant Protein purification |
| Includes | CD-ROM, which contains the Firmware, installation software and Technical Manual. |
| Product Type | Firmware |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.