Description
- Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
- Standard Elution Volume (SEV) is 200-400μL
- Low Elution Volume (LEV) is 30-100μL
- Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Product Type | Cartridge Rack |
| Quantity | 1/Ea. |