Learn More
Promega Maxwell™ 16 LEV Magnet
Catalog No. PRAS1261 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
PRAS1261 1/Ea.
1 options
Catalog No. PRAS1261 Supplier Promega Supplier No. AS1261
Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200-400μL
  • Low Elution Volume (LEV) is 30-100μL
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Specifications
Content And Storage 15°C to 30°C
Product Type Magnet
Quantity 1/Ea.