Description
- No detectable contaminating genomic DNA means fewer repeated experiments or unexplained variable results
- Efficient processing, superior sample capacity
- For most sample types, eluted RNA concentration is directly ready for QRT-PCR; no post-purification concentration required
Maxwell 16 Total RNA Purification Kit
- Extracts total RNA from white blood cell fraction of whole blood, tissue culture cell lines
- PAXgene™-stabilized whole blood and plant leaf tissue can be used in RNA cleanup applications from TRIZOL™ extractions or in vitro transcription reactions
- β-Mercaptoethanol is included for use with certain sample types
- Provide high-concentration (100ng/μL), essentially DNA-free total RNA from cultured cells, mammalian tissue and PAXgene-stabilized white blood cells
- It's personal automation working for you, so you can get more consistent gene expression analysis results with less hands-on labor
Specifications
Specifications
| For Use With (Application) | Automated total RNA purification, qRT-PCR, RT-PCR and cDNA synthesis |
| Includes | Maxwell™ 16 LEV total RNA cartridges 48, RNA Lysis Buffer (RLA) 50mL, RNA dilution Buffer (RDB) 44mL, 97.4% beta-mercaptoethanol (BME) 0.9mL, CytoSol Buffer 10mL, Nuclease-free Water 25mL, LEV plungers 50 , 50 elution tubes (0.5mL) |
| Product Type | Cell LEV Total RNA Kit |
| Quantity | 48 Preps |
For Research Use Only (RUO)