Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell™ CSC Blood DNA Kit

Catalog No. PRAS1321 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1321 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1321 Supplier Promega Supplier No. AS1321
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell CSC Blood DNA Kit is intended for use as an in vitro diagnostic medical device, in combination with the Maxwell CSC Instrument and blood DNA purification method, to perform automated isolation of genomic DNA from human whole blood samples.

  • Automated Genomic DNA Isolation from Blood for In Vitro Diagnostic Assays
  • Purifies high-quality DNA from 1-16 whole blood samples
  • Intended for in vitro diagnostic use in combination with the Maxwell(R) CSC Instrument
  • Offers clinical customers a high-quality DNA extraction method manufactured under cGMP

Specifications

Content And Storage 15°C to 30°C
Format Kit
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.