Learn More
Promega Maxwell™ CSC DNA FFPE Kit
Catalog No. pras1350 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
48 Preps
Catalog No. Quantity
PRAS1350 48 Preps
1 options
Catalog No. PRAS1350 Supplier Promega Supplier No. AS1350
Only null left
Add to Cart
Add to Cart
The Maxwell CSC DNA FFPE Kit is intended for use as an in vitro diagnostic (IVD) medical device to perform automated isolation of DNA from human breast, lung and colon FFPE (formalin-fixed, paraffin-embedded) tissue samples.
  • Automated DNA Purification from FFPE Samples for In Vitro Diagnostic Assays
  • Purifies human DNA from formalin-fixed, paraffin-embedded (FFPE) colon, breast and lung tissues
  • Purifies high-quality DNA suitable for use in amplification-based in vitro diagnostic assays
  • Offers clinical customers a high-quality DNA extraction method manufactured under cGMP
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps