Learn More
Promega Maxwell CSC Installation Qualification, 1 each
Catalog No. PRSA1140 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRSA1140 1 Ea.
1 options
Catalog No. PRSA1140 Supplier Promega Supplier No. SA1140
The Installation Qualification product checks the instrument, documents functionality and demonstrates that everything ordered is supplied. The Operational Qualification product demonstrates the instrument functions according to specifications.
  • Choose the Installation and Operational Qualification Products that Best Suit Your Needs
  • The Installation Qualification (IQ) includes a series of instrument checks, delivers written documentation of functionality and demonstrates that everything ordered with the instrument is supplied and installed
  • The Operational Qualification (OQ) service product demonstrates that the instrument functions according to its operational specifications
  • The IQ/OQ Package combines both services
Specifications
Quantity 1 Ea.