Learn More
Promega Maxwell CSC Instrument, 1 each
Catalog No. PRAS6000 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell CSC Instrument, 1 each
Catalog No.
PRAS6000 Maxwell CSC Instrument, 1 each
1 options
This item is not returnable. View return policy
Catalog No. PRAS6000 Supplier Promega Supplier No. AS6000
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
The Maxwell Clinical Sample Concentrator (CSC) Instrument provides automated nucleic acid purification for a range of clinical sample types. Sample lysis and binding to paramagnetic particles comprise the primary separation principle.
  • Nucleic Acid Extraction for IVD Use
  • Designed and manufactured under cGMP
  • Generates consistent, high−quality nucleic acid for use in downstream diagnostic amplification assays
  • Provides many of the required technical elements of a 21 CFR Part 11 compliant system (user authentication and authorization, data integrity and protection, electronic signatures and audit trails) when used with the appropriate laboratory workflow
Specifications
Isolation Technology Magnetic Bead
Product Type Nucleic Acid Extraction Kit