Learn More
Promega Maxwell RSC Instrument, 1 each
Catalog No. pras4500 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRAS4500 1 Ea.
1 options
Catalog No. PRAS4500 Supplier Promega Supplier No. AS4500
Add to Cart
Add to Cart
Request a Quote
The Maxwell Rapid Sample Concentrator (RSC) Instrument is a platform for automated purification of nucleic acid from a range of sample types. Purification methods use sample lysis and binding to paramagnetic particles as the primary separation principle.
  • Automated Nucleic Acid Purification with Integrated Quantitation
  • High−quality nucleic acid purification with minimal steps and less hands−on time
  • Integrated Quantus™ Fluorometer for sample quantitation
  • Intuitive software and touch screen interface
Specifications
Quantity 1 Ea.