Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell™ RSC Blood DNA Kit

Catalog No. PRAS1400 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1400 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1400 Supplier Promega Supplier No. AS1400
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC Blood DNA Kit is designed for optimal automated extraction of DNA from up to 250μL of whole blood samples using the Maxwell RSC Instrument.

  • Automated DNA Purification from Whole Blood
  • Extracts DNA from 50-300μL of whole blood with a simple, walk-away protocol
  • Provides high yields of pure and amplifiable DNA
  • Compatible with blood stored in EDTA, heparin and citrate anticoagulants

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.