Learn More
Promega Maxwell RSC Buccal Swab DNA Kit, 48 preps
Catalog No. PRAS1640 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Buccal Swab DNA Kit, 48 preps
Catalog No.
PRAS1640 Maxwell RSC Buccal Swab DNA Kit, 48 preps
1 options
Catalog No. PRAS1640 Supplier Promega Supplier No. AS1640
The Maxwell RSC Buccal Swab DNA Purification Kit is designed for automated extraction of DNA from buccal swabs using the Maxwell RSC Instrument.
  • Automated DNA Extraction from Buccal Swabs
  • Optimized reagents for buccal swab extraction
  • Decreased hands−on time with simple protocol
  • Consistent results with sufficient DNA for HLA assays
Specifications
Format Kit
Isolation Technology Magnetic Bead
Product Type DNA Purification Kit