Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC Buffy Coat DNA Kit, 48 preps

Catalog No. PRAS1540 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Buffy Coat DNA Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1540 Maxwell RSC Buffy Coat DNA Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1540 Supplier Promega Supplier No. AS1540
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC Buffy Coat DNA Kit is a simple, automated method for extraction of genomic DNA from 50-250ul of buffy coat samples using the Maxwell RSC Instrument.

  • Automated DNA Purification from Buffy Coat samples
  • Purifies high yields of DNA from 50−250μl of buffy coat samples in about 50 minutes
  • Simple walk−away protocol with no preprocessing
  • Compatible with blood stored in EDTA, heparin and citrate anticoagulants

Specifications

Content And Storage 15°C to 30°C
Format Kit
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.