Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC ccfDNA Plasma Kit, 48 preps

Catalog No. PRAS1480 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC ccfDNA Plasma Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1480 Maxwell RSC ccfDNA Plasma Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1480 Supplier Promega Supplier No. AS1480
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC ccfDNA Plasma Kit is used with the Maxwell RSC Instrument to provide an easy method for efficient, automated purification of circulating cell-free DNA (ccfDNA) from human plasma samples.

  • Automated ccfDNA Purification with a Simple Three−Step Protocol
  • Purifies circulating cell−free DNA from up to 1ml of plasma
  • Consistently high yields of amplifiable ccfDNA
  • Simple, three−step protocol with no preprocessing

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 48 Preps
Product Type ccfDNA Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.