Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC Cultured Cells DNA Kit, 48 preps

Catalog No. PRAS1620 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Cultured Cells DNA Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1620 Maxwell RSC Cultured Cells DNA Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1620 Supplier Promega Supplier No. AS1620
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Maxwell RSC Cultured Cells DNA Kit is used with the Maxwell RSC Instrument to provide a simple method for efficient, automated extraction of DNA from up to 5 x 10^6 mammalian tissue culture cells and 2 x 10^9 bacterial cells.

  • Automated DNA Purification from Mammalian and Bacterial Cultured Cells
  • Extracts DNA from up to 5 x 106 mammalian tissue culture cells and 2 x 109 bacterial cells
  • Simple, walk−away protocol requires no sample preprocessing
  • Purified DNA is ready for analysis in about 45 minutes

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Magnetic Bead
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.