Learn More
Promega Corporation Maxwell™ RSC Enviro TNA Kit
Catalog No. PRAS1831 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Type: Detection Kit
Catalog No.
PRAS1831 Type: Detection Kit
1 options
Catalog No. PRAS1831 Supplier Promega Corporation Supplier No. AS1831
Process 40 mL wastewater samples using Direct Capture and Purification of Viral DNA/RNA From Wastewater. Extract TNA in under 2 hours, efficiently removing PCR inhibitors. Available in manual, Maxwell™ automation, or high-throughput formats, including a startup bundle with vacuum manifold and purification kit.
  • Automated DNA extraction from diverse human sample types.
  • Produces high-quality DNA suitable for various assays.
  • Processes multiple sample types simultaneously.
  • Consistent yield and purity across different samples.
  • Streamlined workflow with minimal manual intervention.
  • Compatible with the Maxwell™ RSC Instrument.
  • Ensures reliable and reproducible results.
  • Ideal for clinical and research applications.
  • Reduces the risk of cross-contamination.
  • High-performance DNA extraction for sensitive downstream applications.
Specifications
Format Maxwell Automation
Isolation Technology Spin or Vacuum Column, Silica Spin Column
Product Type Detection Kit