Learn More
Promega Maxwell™ RSC miRNA Tissue Kit
Catalog No. pras1460 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
48 Preps
Catalog No. Quantity
PRAS1460 48 Preps
1 options
Catalog No. PRAS1460 Supplier Promega Supplier No. AS1460
Only null left
Add to Cart
Add to Cart
The Maxwell(R) RSC miRNA Kit, in combination with the Maxwell(R) RSC Instrument, purifies high-quality total RNA with an enhanced miRNA enrichment.
  • Automated Purification of High-Quality Total RNA with an Enhanced miRNA Enrichment
  • High-quality, amplifiable RNA from mammalian tissue samples
  • Simple, safe RNA extraction without organic reagents
  • Automated RNA extraction of 1-48 samples in a single run
Specifications
Content And Storage 15°C to 30°C
Quantity 48 Preps