Learn More
Promega Maxwell RSC Operational Qualification, 1 each
Catalog No. PRSA1348 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
PRSA1348 1 Ea.
1 options
Catalog No. PRSA1348 Supplier Promega Supplier No. SA1348
Add to Cart
Add to Cart
Request a Quote
Maxwell RSC Installation Qualification and Operational Qualification Products
  • Choose the Installation and Operational Qualification Products that Best Suit Your Needs
  • The Installation Qualification (IQ) includes a series of instrument checks, delivers written documentation of functionality and demonstrates that everything ordered with the instrument is supplied and installed
  • The Operational Qualification (OQ) service product demonstrates that the instrument functions according to its operational specifications
  • The IQ/OQ Package combines both services
Specifications
Quantity 1 Ea.