Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell RSC Plant DNA Kit, 48 preps

Catalog No. PRAS1490 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Maxwell RSC Plant DNA Kit, 48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1490 Maxwell RSC Plant DNA Kit, 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1490 Supplier Promega Supplier No. AS1490
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The Maxwell RSC Plant DNA Kit is used with the Maxwell RSC Instrument (Cat.# AS4500) to provide an easy method for efficient, automated purification of genomic DNA (gDNA) from plant tissue samples.

  • Automated DNA Extraction from Plant Tissues
  • Extracts DNA from a range of plant tissues, including soybean, corn and Arabidopsis
  • Consistent purification, no organic extractions and minimal preprocessing
  • Purified DNA is ready to use in downstream applications including amplification assays

Specifications

Content And Storage 15°C to 30°C
Format Kit
Isolation Technology Organic Extraction
No. of Reactions 48 Preps
Product Type DNA Purification Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.