Learn More
Description
- Automated RNA Extraction from Plant Tissues
- Extracts RNA from a range of plant sample types with no organic reagents
- Cellulose−based paramagnetic particles offer higher binding capacity for increased yields
- Extracted RNA is ready for downstream applications
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Kit |
| Isolation Technology | Magnetic Bead |
| No. of Reactions | 48 Preps |
| Product Type | RNA Extraction Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.